Tisk…

Anti-CFLAR Antibody Pair

Dodavatel: Abnova
ABNOH00008837-AP51EA 23090 CZK
ABNOH00008837-AP51
Anti-CFLAR Antibody Pair
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human CFLAR.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-CFLAR, IgG2a kappa (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-CFLAR (50 µg);

Type:
Antigen: CFLAR
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR