Tisk…

Anti-HMGB1 Mouse Monoclonal Antibody (Biotin)

Dodavatel: Abnova
ABNOH00003146-AP41EA 23090 CZK
ABNOH00003146-AP41
Anti-HMGB1 Mouse Monoclonal Antibody (Biotin)
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human HMGB1.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-HMGB1, IgG2a kappa (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-HMGB1, IgG1 kappa (50 ?g);

Type:
Antigen: HMGB1
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR