Tisk…

Anti-S100 A7 Mouse Monoclonal Antibody (Biotin)

Dodavatel: Abnova
ABNOH00006278-AP41EA 23800 CZK
ABNOH00006278-AP41
Anti-S100 A7 Mouse Monoclonal Antibody (Biotin)
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human S100A7.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-S100A7, IgG1 kappa (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-S100A7, IgG2a kappa (50 ?g);

Type:
Antigen: S100 A7
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR