Tisk…
Anti-RETN Mouse Monoclonal Antibody (Biotin)
Dodavatel: Abnova
ABNOH00056729-AP41EA 23090 CZK
ABNOH00056729-AP41
Anti-RETN Mouse Monoclonal Antibody (Biotin)
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human RETN.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-RETN, IgG2a kappa (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-RETN, IgG3 kappa (50 ?g);
Type:
Antigen: RETN
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human