Tisk…

Anti-F2 Antibody Pair

Dodavatel: Abnova
ABNOH00002147-AP52EA 23090 CZK
ABNOH00002147-AP52
Anti-F2 Antibody Pair
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human F2.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-F2, IgG Mix kappa (300 µg); 2. Detection Antibody: Rabbit Polyclonal Anti-F2 (50 µl);

Type:
Antigen: F2
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR