Tisk…

Anti-ART3 Polyclonal Antibody Pair

Dodavatel: Abnova
ABNOH00000419-PW3EA 23800 CZK
ABNOH00000419-PW3
Anti-ART3 Polyclonal Antibody Pair
Protilátky
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-ART3 (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-ART3 (50 µg)

Type:
Antigen: ART3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR