Tisk…

Anti-TNFRSF19 Mouse Monoclonal Antibody (Biotin)

Dodavatel: Abnova
ABNOH00055504-AP42EA 23090 CZK
ABNOH00055504-AP42
Anti-TNFRSF19 Mouse Monoclonal Antibody (Biotin)
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human TNFRSF19.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-TNFRSF19, IgG1 kappa (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-TNFRSF19, IgG2b kappa (50 ?g);

Type:
Antigen: TNFRSF19
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR