Tisk…
Anti-PIR Polyclonal Antibody Pair
Dodavatel: Abnova
ABNOH00008544-AP22EA 23800 CZK
ABNOH00008544-AP22
Anti-PIR Polyclonal Antibody Pair
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human PIR.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-PIR (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-PIR (20 µg);
Type:
Antigen: PIR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human