Tisk…
Anti-CDX4 Mouse Monoclonal Antibody (Biotin)
Dodavatel: Abnova
ABNOH00001046-AP41EA 23800 CZK
ABNOH00001046-AP41
Anti-CDX4 Mouse Monoclonal Antibody (Biotin)
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human CDX4.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-CDX4, IgG2a kappa (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-CDX4, IgG2a kappa (50 ?g);
Type:
Antigen: CDX4
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human