Tisk…
Anti-ERCC8 Polyclonal Antibody Pair
Dodavatel: Abnova
ABNOH00001161-PW2EA 23090 CZK
ABNOH00001161-PW2
Anti-ERCC8 Polyclonal Antibody Pair
Protilátky
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-ERCC8 (300 ?l); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-ERCC8 (50 ?g)