Tisk…

Anti-ORM1 Antibody Pair

Dodavatel: Abnova
ABNOH00005004-AP61EA 23090 CZK
ABNOH00005004-AP61
Anti-ORM1 Antibody Pair
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human ORM1.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-ORM1, IgG1 kappa (100 µg); 2. Detection Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-ORM1 (50 µg);

Type:
Antigen: ORM1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR