Tisk…
Anti-GYG2 Polyclonal Antibody Pair
Dodavatel: Abnova
ABNOH00008908-AP22EA 23090 CZK
ABNOH00008908-AP22
Anti-GYG2 Polyclonal Antibody Pair
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human GYG2.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-GYG2 (100 ?g); 2. Detection Antibody: Mouse purified Polyclonal Anti-GYG2 (20 ?g);