Tisk…

Anti-Bcl10 Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPB9525EA 13000 CZK
BSBTPB9525
Anti-Bcl10 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for B-cell lymphoma/leukemia 10(BCL10) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: Bcl10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR