Tisk…

Anti-cIAP2 Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPB9527EA 13000 CZK
BSBTPB9527
Anti-cIAP2 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Baculoviral IAP repeat-containing protein 3(BIRC3) detection. Tested with WB in Human.

Type: Primary
Antigen: cIAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR