Tisk…

Anti-BMPR1B Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPB9528EA 14410 CZK
BSBTPB9528
Anti-BMPR1B Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Bone morphogenetic protein receptor type-1B(BMPR1B) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: BMPR1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR