Tisk…

Anti-CA1 Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPB9529EA 14410 CZK
BSBTPB9529
Anti-CA1 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Carbonic anhydrase 1(CA1) detection. Tested with WB, IHC-P in Human;Rat.

Type: Primary
Antigen: CA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR