Tisk…

Anti-Cdc6 Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPB9531EA 14410 CZK
BSBTPB9531
Anti-Cdc6 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Cell division control protein 6 homolog(CDC6) detection. Tested with WB, IHC-P in Human;Rat.

Type: Primary
Antigen: Cdc6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR