Tisk…

Anti-Cdk2 Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPB9534EA 14410 CZK
BSBTPB9534
Anti-Cdk2 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Cyclin-dependent kinase 2(CDK2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: Cdk2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR