Tisk…

Anti-Cdk7 Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPB9536EA 16320 CZK
BSBTPB9536
Anti-Cdk7 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Cyclin-dependent kinase 7(CDK7) detection. Tested with WB in Human.

Type: Primary
Antigen: Cdk7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR