Tisk…
Anti-Cryptochrome I Rabbit Polyclonal Antibody
Dodavatel: Boster
BSBTPB9540EA 15510 CZK
BSBTPB9540
Anti-Cryptochrome I Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Cryptochrome-1(CRY1) detection. Tested with WB in Human;Rat.
Type: Primary
Antigen: Cryptochrome I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat