Tisk…

Anti-CYLD Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPB9543EA 13000 CZK
BSBTPB9543
Anti-CYLD Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Ubiquitin carboxyl-terminal hydrolase CYLD(CYLD) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: CYLD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR