Tisk…

Anti-CYP24A1 Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPB9547EA 15510 CZK
BSBTPB9547
Anti-CYP24A1 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for 1,25-dihydroxyvitamin D(3) 24-hydroxylase, mitochondrial (CYP24A1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: CYP24A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR