Tisk…

Anti-CCN1 Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPB9549EA 16320 CZK
BSBTPB9549
Anti-CCN1 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Protein CYR61(CYR61) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: CCN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR