Tisk…

Anti-MCSF Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPB9562EA 14410 CZK
BSBTPB9562
Anti-MCSF Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Macrophage colony-stimulating factor 1(CSF1) detection. Tested with IHC-P, ELISA in Mouse;Rat.

Type: Primary
Antigen: MCSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR