Tisk…

Anti-Cystatin A Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPB9564EA 14410 CZK
BSBTPB9564
Anti-Cystatin A Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Cystatin-A(CSTA) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: Cystatin A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR