Tisk…
VWR®, Culture tubes, round bottom, disposable
Dodavatel: VWR
VWR®
47729-576EA 0 CZK
VWRU47729-566 VWRU47729-586 VWRU47729-574 VWRU47729-584 VWRU47729-583 47729-572 VWRU47729-570 VWRU47729-580 47729-576 47729-568 VWRU47729-578
VWR®, Culture tubes, round bottom, disposable
Zkumavky Zkumavky kultivační
Manufactured from type I, class B borosilicate glass.
  • Chemically inert
  • High heat tolerance
  • Uniform dimensions
  • Convenient dispensing packages
Eeach tube is heat-treated at temperatures exceeding 500 °C (932°F) immediately before packing, which eliminates contamination, increases strength, and reduces breakage. VWR® disposable culture tubes are resistant to chemical attack and weathering. Contents will not leach out and contaminate solutions. Borosilicate glass construction prevents pH change in laboratory tests. Tubes feature well-rounded bottoms and fire-polished tops free of rough edges. Dimensional specifications of these tubes are rigidly controlled for reliable, uniform results. Tubes are clean-sealed in a convenient dispensing package to keep them free from dust and other particulate contamination.
Certifikace: Tubes exceed USP Type I specifications.
Each tube is heat-treated at temperatures exceeding 500 °C (932 °F) immediately before packing, which eliminates contamination, increases strength, and reduces breakage. VWR® disposable culture tubes are resistant to chemical attack and weathering. Contents will not leach out and contaminate solutions. Borosilicate glass construction prevents pH change in laboratory tests. Tubes feature well-rounded bottoms and fire-polished tops free of rough edges. Dimensional specifications of these tubes are rigidly controlled for reliable, uniform results. Tubes are clean-sealed in a convenient dispensing package to keep them free from dust and other particulate contamination.
Certifikace: Tubes exceed USP Type I specifications.