Order Entry
SSL Broth and OFS Agar
SSL Broth and OFS Agar
Supplier:  DOEHLER GROUP
Certificates
undefined
SSL Broth and OFS Agar
Supplier:  DOEHLER GROUP
Certificates
Related Information
Product Information
  • Catalog #
    Supplier #
    Standards
    Filling
    Pack type
    Pk
    Availability
    Price
  • OFS-agar
    OFS-agar
    710-0739
    204707782
    -
    250 ml
    Bottle
    9
    Log in for availability
    Log in for availability
    1 * 9 KS
    1 * 9 KS - 1 360,00 Kč
    1 360,00 Kč
    1 * 9 KS
       1 360,00 Kč
  • SSL-broth, 250 ml
    SSL-broth, 250 ml
    710-0852
    204712782
    -
    250 ml
    Bottle
    9
    Log in for availability
    Log in for availability
    1 * 9 KS
    1 * 9 KS - 1 610,00 Kč
    1 610,00 Kč
    1 * 9 KS
       1 610,00 Kč