Tisk…
Test sieves, 300×75 mm
Dodavatel: Endecotts
510-0313EA 9350 CZK
510-0313 ENDE300BIW12.5
Test sieves, 300×75 mm
Síta Síta
Brass woven wire mesh test sieves are ideal for all types of laboratory sampling and particle size analysis.
For more size options please contact your local Avantor office.
Certifikace: ISO3310-1:2000, BS410-1:2000; DIN-ISO3310-1; NF ISO3310-1:2000
Informace o dodávce: Each sieve is supplied together with a certificate of examination and carries the BS 410 kite mark.