Tisk…
Test sieves, 100×40 mm
Dodavatel: Endecotts
510-0213EA 0 CZK
ENDE100BIW.710 ENDE100BIW16.0 ENDE100BIW.053 ENDE100BIW8.00 ENDE100BIW2.80 ENDE100BIW9.50 ENDE100BIW.090 ENDE100BIW1.70 ENDE100BIW1.40 ENDE100BIW4.75 ENDE100BIW.250 ENDE100BIW2.36 ENDE100BIW11.2 ENDE100BIW.850 ENDE100BIW.212 ENDE100BIW.355 ENDE100BIW13.2 ENDE100BIW.038 ENDE100BIW.600 ENDE100BIW.063 ENDE100BIW6.70 ENDE100BIW5.60 ENDE100BIW4.00 ENDE100BIW3.35 ENDE100BIW1.18 ENDE100BIW.106 ENDE100BIW.300
Test sieves, 100×40 mm
Brass woven wire mesh test sieves are ideal for all types of laboratory sampling and particle size analysis.
For more size options please contact your local Avantor office.
Certifikace: ISO3310-1:2000, BS410-1:2000; DIN-ISO3310-1; NF ISO3310-1:2000
Informace o dodávce: Each sieve is supplied together with a certificate of examination and carries the BS 410 kite mark.