Tisk…
Filter discs, biplane, series 16, porosity 5, VitraPOR®
Dodavatel: ROBU
VitraPOR®
ROBU16905EA 15780 CZK
ROBU16905 ROBU16805 ROBU16125 ROBU16255 ROBU16305 ROBU16205 ROBU16105 ROBU16115 ROBU16705 ROBU16605 ROBU16505 ROBU16405
Filter discs, biplane, series 16, porosity 5, VitraPOR®
Filtry Disky filtrační
The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
  • Very good chemical resistance
  • Very good temperature resistance
Made of borosilicate glass. The faces of filter discs from the series 16 are precisely ground down to tight tolerances and plane surfaces as may be required in specific installation circumstances.