Tisk…

X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt), peqGOLD

Dodavatel: VWR Peqlab

PEQLAB peqGOLD
PEQL37-2725EA 0 CZK
PEQL37-2725 730-1501 730-1500 730-1499
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt), peqGOLD
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt)

Substrate for the detection of beta-glucuronidase.

Vzorec: C₂₀H₂₆BrClN₂O₇
MW: 521,79 g/mol
MDL Number: MFCD00058543
CAS číslo: 114162-64-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR