Tisk…

Anti-AHSG Antibody

Dodavatel: Boster
BSBTPB9568EA 15510 CZK
BSBTPB9568
Anti-AHSG Antibody
Protilátky
Rabbit IgG polyclonal antibody for Alpha-2-HS-glycoprotein(AHSG) detection. Tested with WB, IHC-P, ELISA in Human;Rat.

Type: Primary
Antigen: AHSG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR