Tisk…

Anti-Eotaxin Antibody

Dodavatel: Boster
BSBTPB9569EA 13000 CZK
BSBTPB9569
Anti-Eotaxin Antibody
Protilátky
Rabbit IgG polyclonal antibody for Eotaxin(CCL11) detection. Tested with WB, ELISA in Human.

Type: Primary
Antigen: Eotaxin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR