Tisk…

Anti-RANTES Antibody

Dodavatel: Boster
BSBTPB9573EA 13000 CZK
BSBTPB9573
Anti-RANTES Antibody
Protilátky
Rabbit IgG polyclonal antibody for C-C motif chemokine 5(CCL5) detection. Tested with WB, ELISA in Human.

Type: Primary
Antigen: RANTES
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR