Tisk…

Anti-DLD Antibody

Dodavatel: Boster
BSBTPB9579EA 14410 CZK
BSBTPB9579
Anti-DLD Antibody
Protilátky
Rabbit IgG polyclonal antibody for Dihydrolipoyl dehydrogenase, mitochondrial(DLD) detection. Tested with WB, IHC-P in Human;Mouse; Rat.

Type: Primary
Antigen: DLD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR