Tisk…

Anti-EIF6 Antibody

Dodavatel: Boster
BSBTPB9582EA 14410 CZK
BSBTPB9582
Anti-EIF6 Antibody
Protilátky
Rabbit IgG polyclonal antibody for Eukaryotic translation initiation factor 6(EIF6) detection. Tested with WB, IHC-P in Human;Mouse; Rat.

Type: Primary
Antigen: EIF6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR