Tisk…

Anti-Eph receptor B1 Antibody

Dodavatel: Boster
BSBTPB9584EA 14410 CZK
BSBTPB9584
Anti-Eph receptor B1 Antibody
Protilátky
Rabbit IgG polyclonal antibody for Ephrin type-B receptor 1(EPHB1) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: Eph receptor B1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR