Tisk…

Anti-Fascin Antibody

Dodavatel: Boster
BSBTPB9592EA 13000 CZK
BSBTPB9592
Anti-Fascin Antibody
Protilátky
Rabbit IgG polyclonal antibody for Fascin(FSCN1) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: Fascin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR