Tisk…

Anti-Frizzled homolog 1 Antibody

Dodavatel: Boster
BSBTPB9594EA 15510 CZK
BSBTPB9594
Anti-Frizzled homolog 1 Antibody
Protilátky
Rabbit IgG polyclonal antibody for Frizzled-1(FZD1) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: Frizzled homolog 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR