Tisk…

Anti-GJA3 Antibody

Dodavatel: Boster
BSBTPB9596EA 16320 CZK
BSBTPB9596
Anti-GJA3 Antibody
Protilátky
Rabbit IgG polyclonal antibody for Gap junction alpha-3 protein(GJA3) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: GJA3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR