Tisk…

Anti-IDH1 Antibody

Dodavatel: Boster
BSBTPB9601EA 14410 CZK
BSBTPB9601
Anti-IDH1 Antibody
Protilátky
Rabbit IgG polyclonal antibody for Isocitrate dehydrogenase [NADP] cytoplasmic(IDH1) detection. Tested with WB, IHC-P in Human;Mouse; Rat.

Type: Primary
Antigen: IDH1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR