Tisk…

Anti-IDO1 Antibody

Dodavatel: Boster
BSBTPB9603EA 14410 CZK
BSBTPB9603
Anti-IDO1 Antibody
Protilátky
Rabbit IgG polyclonal antibody for Indoleamine 2,3-dioxygenase 1(IDO1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: IDO1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR