Tisk…

Anti-Lipocalin 2 Antibody

Dodavatel: Boster
BSBTPB9608EA 14410 CZK
BSBTPB9608
Anti-Lipocalin 2 Antibody
Protilátky
Rabbit IgG polyclonal antibody for Neutrophil gelatinase-associated lipocalin(LCN2) detection. Tested with WB, IHC-P, ELISA in Mouse; Rat.

Type: Primary
Antigen: Lipocalin 2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR