Tisk…

Anti-MICA Antibody

Dodavatel: Boster
BSBTPB9612EA 16320 CZK
BSBTPB9612
Anti-MICA Antibody
Protilátky
Rabbit IgG polyclonal antibody for MHC class I polypeptide-related sequence A(MICA) detection. Tested with WB in Human.

Type: Primary
Antigen: MICA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR