Tisk…

Anti-MNAT1 Antibody

Dodavatel: Boster
BSBTPB9614EA 15510 CZK
BSBTPB9614
Anti-MNAT1 Antibody
Protilátky
Rabbit IgG polyclonal antibody for CDK-activating kinase assembly factor MAT1(MNAT1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: MNAT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR