Tisk…

Anti-p95 NBS1 Antibody

Dodavatel: Boster
BSBTPB9615EA 14410 CZK
BSBTPB9615
Anti-p95 NBS1 Antibody
Protilátky
Rabbit IgG polyclonal antibody for Nibrin(NBN) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: p95 NBS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR