Tisk…

Anti-Wnt-4 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3574R-100EA 7780 CZK
BIOV3574R-100
Anti-Wnt-4 Rabbit Polyclonal Antibody
Wnt gene family members, including Wnt-1 throμgh Wnt-10, play a key role in regulating cellular growth and differentiation. Wnt-1 is a cysteine-rich, secreted glycoprotein that associates with cell membranes and likely functions as a key regulator of cellular adhesion. Wnt-3 is also involved in tumorigenesis and Wnt-2 and Wnt-4 may be associated with abnormal proliferation in human breast tissue.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: Wnt-4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR