Tisk…

Anti-Oct-2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3575-100EA 7470 CZK
BIOV3575-100
Anti-Oct-2 Rabbit Polyclonal Antibody
Protilátky
Members of Oct family of transcription factors specifically interact with Octamer motif ATGCAAAT, a regulatory element important for tissue- and cell-specific transcription as well as for transcription of a number of housekeeping genes. All of the members of the Oct family contain two highly conserved domains which are separated by 14-26 variable amino acids. These include the POU homeodomain and the POU-specific domain. Both are required for DNA binding and are involved in protein-protein interactions. Evidences indicate that regulation of Oct family transcription factors occurs at the level of phosphorylation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Oct-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR