Tisk…

Anti-BCMA Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3577-100EA 7470 CZK
BIOV3577-100
Anti-BCMA Rabbit Polyclonal Antibody
Protilátky
BCMA is a recently identified receptor for BAFF. It binds BAFF and April. BAFF and its receptors induce B cell development and survival, activate NF-kB and c-Jun N-terminal kinase, and are involved in B cell associated autoimmune diseases.

Recommended Dilutions: Western Blot: 2-6 µg/ml; Immunohistochemistry: 5-20 µg/ml

Type: Primary
Antigen: BCMA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR