Tisk…

Anti-BAFF-R Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3578R-100EA 6910 CZK
BIOV3578R-100
Anti-BAFF-R Rabbit Polyclonal Antibody
Protilátky
Members in the TNF superfamily regulate immune responses and induce apoptosis. A novel member in the TNF family was recently identified by several groups and designated BAFF, BLyS, TALL-1, THANK, and zTNF4. BAFF/BLyS was characterized as a B cell activator since it induced B cell proliferation and immunoglobulin secretion. Two receptors, TACI and BCMA, for BAFF were originally identified. A third receptor was identified recently and designated BAFF-R and BR3 for BLyS receptor 3. Unlike BCMA and TACI, which bind to BAFF and April, BAFF-R/BR3 is specific for BAFF and plays a predominant role in BAFF induced B cell development and survival. BAFF and its receptors are involved in B cell associated autoimmune diseases, and activate NF-kB and c-Jun N-terminal kinase.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BAFF-R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR